Merck
  • Lower fetuin-A, retinol binding protein 4 and several metabolites after gastric bypass compared to sleeve gastrectomy in patients with type 2 diabetes.
Lower fetuin-A, retinol binding protein 4 and several metabolites after gastric bypass compared to sleeve gastrectomy in patients with type 2 diabetes.
PloS one (2014-05-08)
Mia Jüllig, Shelley Yip, Aimin Xu, Greg Smith, Martin Middleditch, Michael Booth, Richard Babor, Grant Beban, Rinki Murphy
ABSTRACT
Bypass of foregut secreted factors promoting insulin resistance is hypothesized to be one of the mechanisms by which resolution of type 2 diabetes (T2D) follows roux-en-y gastric bypass (GBP) surgery. To identify insulin resistance-associated proteins and metabolites which decrease more after GBP than after sleeve gastrectomy (SG) prior to diabetes remission. Fasting plasma from 15 subjects with T2D undergoing GBP or SG was analyzed by proteomic and metabolomic methods 3 days before and 3 days after surgery. Subjects were matched for age, BMI, metformin therapy and glycemic control. Insulin resistance was calculated using homeostasis model assessment (HOMA-IR). For proteomics, samples were depleted of abundant plasma proteins, digested with trypsin and labeled with iTRAQ isobaric tags prior to liquid chromatography-tandem mass spectrometry analysis. Metabolomic analysis was performed using gas chromatography-mass spectrometry. The effect of the respective bariatric surgery on identified proteins and metabolites was evaluated using two-way analysis of variance and appropriate post-hoc tests. HOMA-IR improved, albeit not significantly, in both groups after surgery. Proteomic analysis yielded seven proteins which decreased significantly after GBP only, including Fetuin-A and Retinol binding protein 4, both previously linked to insulin resistance. Significant decrease in Fetuin-A and Retinol binding protein 4 after GBP was confirmed using ELISA and immunoassay. Metabolomic analysis identified significant decrease of citrate, proline, histidine and decanoic acid specifically after GBP. Greater early decrease was seen for Fetuin-A, Retinol binding protein 4, and several metabolites after GBP compared to SG, preceding significant weight loss. This may contribute to enhanced T2D remission observed following foregut bypass procedures.
Sigma-Aldrich
Methyl chloroformate, 99%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Iodoacetamide, Vetec, reagent grade, 98%
SAFC
Iodoacetamide