Merck
  • A comparison of cannabidiolic acid with other treatments for anticipatory nausea using a rat model of contextually elicited conditioned gaping.
A comparison of cannabidiolic acid with other treatments for anticipatory nausea using a rat model of contextually elicited conditioned gaping.
Psychopharmacology (2014-03-07)
Erin M Rock, Cheryl L Limebeer, Roshan Navaratnam, Martin A Sticht, Natasha Bonner, Kristin Engeland, Rachel Downey, Heather Morris, Meagan Jackson, Linda A Parker
ABSTRACT
The effectiveness of cannabidiolic acid (CBDA) was compared with other potential treatments for anticipatory nausea (AN), using a rat model of contextually elicited conditioned gaping reactions. The potential of ondansetron (OND), Δ(9)-tetrahydrocannabinol (THC), chlordiazepoxide (CDP), CBDA, and co-administration of CBDA and tetrahydrocannabinolic acid (THCA) to reduce AN and modify locomotor activity was evaluated. Following four pairings of a novel context with lithium chloride (LiCl), the rats were given a test for AN. On the test trial, they received pretreatment injections of the following: vehicle, OND (0.1 or 1.0 mg/kg), THC (0.5 mg/kg), CBDA (0.0001, 0.001, 0.01, 0.1 mg/kg or 1.0 mg/kg), CDP (1, 5, or 10 mg/kg) or co-administration of subthreshold doses of CBDA (0.1 μg/kg), and THCA (5 μg/kg). Immediately following the AN test trial in all experiments, rats were given a 15 min locomotor activity test. Finally, the potential of CBDA (0.001, 0.01, 0.1, and 1 mg/kg) to attenuate conditioned freezing to a shock-paired tone was assessed. THC, CBDA, and CDP, but not OND, reduced contextually elicited gaping reactions. Co-administration of subthreshold doses of CBDA and THCA also suppressed AN, and this effect was blocked by pretreatment with either a cannabinoid receptor 1 (CB1) receptor antagonist or a 5-hydroxytryptamine 1A (5-HT1A) receptor antagonist. CDP (but not CBDA, THC or CBDA and THCA) also suppressed locomotor activity at effective doses. CBDA did not modify the expression of conditioned fear. CBDA has therapeutic potential as a highly potent and selective treatment for AN without psychoactive or locomotor effects.
Product Description
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, Vetec, reagent grade
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Electrolyte solution, nonaqueous, LiCl in ethanol (saturated)
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)