Merck
Studies of the humoral factors produced by layered chondrocyte sheets.
Journal of tissue engineering and regenerative medicine (2012-11-21)
K Hamahashi, M Sato, M Yamato, M Kokubo, G Mitani, S Ito, T Nagai, G Ebihara, T Kutsuna, T Okano, J Mochida
ABSTRACT
The authors aimed to repair and regenerate articular cartilage with layered chondrocyte sheets, produced using temperature-responsive culture dishes. The purpose of this study was to investigate the humoral factors produced by layered chondrocyte sheets. Articular chondrocytes and synovial cells were harvested during total knee arthroplasty. After co-culture, the samples were divided into three groups: a monolayer, 7 day culture sheet group (group M); a triple-layered, 7 day culture sheet group (group L); and a monolayer culture group with a cell count identical to that of group L (group C). The secretion of collagen type 1 (COL1), collagen type 2 (COL2), matrix metalloproteinase-13 (MMP13), transforming growth factor-β (TGFβ), melanoma inhibitory activity (MIA) and prostaglandin E2 (PGE2) were measured by enzyme-linked immunosorbent assay (ELISA). Layered chondrocyte sheets produced the most humoral factors. PGE2 expression declined over time in group C but was significantly higher in groups M and L. TGFβ expression was low in group C but was significantly higher in groups M and L (p<0.05). Our results suggest that the humoral factors produced by layered chondrocyte sheets may contribute to cartilaginous tissue repair and regeneration.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, suitable for fluorescence, BioReagent, ≥97.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
5-(N-Methyl-N-isobutyl)­amiloride, ≥98% (TLC), powder
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, Fluorogenic reagent
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Dinoprostone, European Pharmacopoeia (EP) Reference Standard