Merck
  • One-step exfoliation and fluorination of boron nitride nanosheets and a study of their magnetic properties.
One-step exfoliation and fluorination of boron nitride nanosheets and a study of their magnetic properties.
Angewandte Chemie (International ed. in English) (2014-03-05)
Miao Du, Xianlei Li, Aizhu Wang, Yongzhong Wu, Xiaopeng Hao, Mingwen Zhao
ABSTRACT
A novel, simple, and efficient method for the preparation of the fluorinated hexagonal boron nitride nanosheets (F-BNNSs) and the corresponding magnetic properties is presented. A one-step route is used to exfoliate and fluorinate the BNNSs by ammonium fluoride (NH4F) from hexagonal boron nitride (h-BN) powder. Through related instrument characterizations and theoretical calculations, we confirm that large-area and few-layer F-BNNSs were successfully produced by this method, which can be attributed to a fluorination-assisted exfoliation mechanism from the bulk h-BN in NH4F. More intriguingly, we initially verified that the as-prepared F-BNNSs exhibit ferromagnetic characteristics, which would have good potential applications in spintronic devices.
Sigma-Aldrich
Boron nitride, powder, ~1 μm, 98%
Sigma-Aldrich
Boron nitride, nanopowder, <150 nm avg. part. size (TEM), 99% trace metals basis