Merck
  • A tocotrienol series with an oxidative terminal prenyl unit from Garcinia amplexicaulis.
A tocotrienol series with an oxidative terminal prenyl unit from Garcinia amplexicaulis.
Phytochemistry (2014-12-04)
Alexis Lavaud, Pascal Richomme, Julia Gatto, Marie-Christine Aumond, Cyril Poullain, Marc Litaudon, Ramaroson Andriantsitohaina, David Guilet
ABSTRACT
Ten tocotrienol derivatives, i.e., amplexichromanols (1-10), were isolated from stem bark of Garcinia amplexicaulis Vieill. ex Pierre collected in Caledonia. The structures of the compounds 1-5 were determined to be chromanol derivatives substituted by a polyprenyl chain oxidized in terminal position. The remaining compounds 6-10 are the corresponding dimeric derivatives. Eleven known compounds, including xanthones, tocotrienol derivatives, triterpenes and phenolic compounds, were also isolated. Their structures were mainly determined using one and two-dimensional NMR and mass spectroscopy analysis. The compounds and some amplexichromanol molecules formerly isolated from G. amplexicaulis exhibited significant antioxidant activity against lipid peroxidation and in the ORAC assay.
Sigma-Aldrich
Methanol-d4, anhydrous, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Methanol-d4, "100%", 99.96 atom % D
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.05 % (v/v) TMS