Merck
  • ⁹⁹mTc-labeled single-domain antibody EG2 in targeting epidermal growth factor receptor: an in-vitro and mouse model in-vivo study.
⁹⁹mTc-labeled single-domain antibody EG2 in targeting epidermal growth factor receptor: an in-vitro and mouse model in-vivo study.
Nuclear medicine communications (2015-01-16)
Chongjiao Li, Bing Wen, Lifei Wang, Hongyan Feng, Xiaotian Xia, Zhiling Ding, Bin Gao, Yongxue Zhang, Xiaoli Lan
ABSTRACT
The aim of this study was to explore the targeting ability and metabolic characteristics of the technetium-99m-labeled single-domain antibody (⁹⁹mTc-sdAb) EG2 targeting epidermal growth factor receptor (EGFR) through an in-vitro and in-vivo study. The sdAb EG2 was radiolabeled with Tc using a tricarbonyl kit. The EGFR expression level of A431 and OCM-1 cells was confirmed using immunofluorescence staining. Cell binding, blocking, uptake, and efflux studies were performed to investigate the binding specificity of ⁹⁹mTc-sdAb EG2 in vitro. Single-photon emission computed tomography imaging and biodistribution studies were used to explore the targeting abilities and metabolic characteristics of Tc-sdAb EG2 in vivo. ⁹⁹mTc-sdAb EG2 was successfully prepared with labeling yields of 60-71% and specific activity of 1.83±0.29 GBq/mg (n=3). Immunofluorescence staining revealed high and low EGFR expression on the surface of A431 and OCM-1 cells, respectively. The binding affinity of ⁹⁹mTc-sdAb EG2 to A431 cells was 43.53±1.89 nmol/l. ⁹⁹mTc-sdAb EG2 uptake in A431 cells in vitro could be blocked by ∼19, 40, and 66% in the presence of excess unlabeled sdAb EG2 at 100, 500, and 1000 nmol/l, respectively. Single-photon emission computed imaging indicated that A431 tumor images could be clearly displayed at early scan time points after ⁹⁹mTc-sdAb EG2 injection, even as early as 1 h. Biodistribution study showed that the A431 tumor uptake of ⁹⁹mTc-sdAb EG2 was blocked by about 51% at 3 h after coinjecting excess of sdAb EG2. However, there were almost no OCM-1 tumor images at the corresponding scan time points and the OCM-1 tumor uptake was only 0.40±0.13% injected dose per gram of tissue (n=5) at 3 h. This study demonstrated that sdAb EG2 can effectively target EGFR in vitro and in vivo in tumors, suggesting that it could be used as a molecular probe for EGFR detection.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Streptomycin sulfate salt, powder
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)