Merck
  • Cyclic tensile strain enhances osteogenesis and angiogenesis in mesenchymal stem cells from osteoporotic donors.
Cyclic tensile strain enhances osteogenesis and angiogenesis in mesenchymal stem cells from osteoporotic donors.
Tissue engineering. Part A (2013-08-10)
Adisri Charoenpanich, Michelle E Wall, Charles J Tucker, Danica M K Andrews, David S Lalush, Douglas R Dirschl, Elizabeth G Loboa
ABSTRACT
We have shown that the uniaxial cyclic tensile strain of magnitude 10% promotes and enhances osteogenesis of human mesenchymal stem cells (hMSC) and human adipose-derived stem cells (hASC) from normal, nonosteoporotic donors. In the present study, MSC from osteoporotic donors were analyzed for changes in mRNA expression in response to 10% uniaxial tensile strain to identify potential mechanisms underlying the use of this mechanical loading paradigm for prevention and treatment of osteoporosis. Human MSC isolated from three female, postmenopausal osteoporotic donors were analyzed for their responses to mechanical loading using microarray analysis of over 47,000 gene probes. Human MSC were seeded in three-dimensional collagen type I constructs to mimic the organic extracellular matrix of bone and 10% uniaxial cyclic tensile strain was applied to promote osteogenesis. Seventy-nine genes were shown to be regulated within hMSC from osteoporotic donors in response to 10% cyclic tensile strain. Upregulation of six genes were further confirmed with real-time RT-PCR: jun D proto-oncogene (JUND) and plasminogen activator, urokinase receptor (PLAUR), two genes identified as potential key molecules from network analysis; phosphoinositide-3-kinase, catalytic, delta polypeptide (PIK3CD) and wingless-type MMTV integration site family, member 5B (WNT5B), two genes with known importance in bone biology; and, PDZ and LIM domain 4 (PDLIM4) and vascular endothelial growth factor A (VEGFA), two genes that we have previously shown are significantly regulated in hASC in response to this mechanical stimulus. Function analysis indicated that 10% cyclic tensile strain induced expression of genes associated with cell movement, cell proliferation, and tissue development, including development in musculoskeletal and cardiovascular systems. Our results demonstrate that hMSC from aged, osteoporotic donors are capable of enhanced osteogenic differentiation in response to 10% cyclic tensile strain with significant increases in the expression of genes associated with enhanced cell proliferation, musculoskeletal development, and angiogenesis. Surprisingly, cyclic tensile strain of magnitude 10% not only enhanced osteogenesis in hMSC from osteoporotic donors, but also enhanced expression of angiogenic factors. Better understanding and methodologies to promote osteogenesis in hMSC from elderly, osteoporotic donors may greatly facilitate achieving long-term success in bone regeneration and functional bone tissue engineering for this ever-growing patient population.
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Phosphatase substrate, powder
Sigma-Aldrich
Phosphatase substrate, 40 mg capsules
Sigma-Aldrich
Phosphatase substrate, 40 mg tablets
Sigma-Aldrich
Phosphatase substrate, 100 mg capsules
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
Sigma-Aldrich
Phosphatase substrate, 5 mg tablets
Sigma-Aldrich
Phosphatase substrate, Suitable for manufacturing of diagnostic kits and reagents
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis