Merck
  • The JAK inhibitor, tofacitinib, reduces the T cell stimulatory capacity of human monocyte-derived dendritic cells.
The JAK inhibitor, tofacitinib, reduces the T cell stimulatory capacity of human monocyte-derived dendritic cells.
Annals of the rheumatic diseases (2013-09-10)
Satoshi Kubo, Kunihiro Yamaoka, Masahiro Kondo, Kaoru Yamagata, Jidong Zhao, Shigeru Iwata, Yoshiya Tanaka
ABSTRACT
Tofacitinib, which is a Janus kinase (JAK) inhibitor, has shown clinical effects in the treatment of rheumatoid arthritis. JAKs are important kinases in lymphocyte differentiation; however, their function in dendritic cells (DCs) is unknown. In this study, the function of JAKs in DCs was investigated with tofacitinib. The effects of tofacitinib on the maturation of human monocyte-derived DCs induced by lipopolysaccharide (LPS) stimulation were investigated. In addition, its effects on T cell stimulatory capability was investigated by coculturing with naïve CD45RA-positive T cells. Tofacitinib decreased expression of CD80/CD86 in a concentration-dependent manner in LPS-stimulated DCs; however, it did not affect HLA-DR expression. Tofacitinib suppressed tumour necrosis factor, interleukin (IL)-6 and IL-1β production without affecting transforming growth factor (TGF)-β and IL-10 production. Meanwhile, CD80/CD86 expression in DCs was enhanced by type I interferon (IFN) stimulation, and the LPS-induced CD80/CD86 expression was inhibited by an antibody to type I IFN receptor. Furthermore, tofacitinib suppressed production of type I IFN and activation of interferon regulatory factor (IRF)-7, which is a transcription factor involved in CD80/CD86 and type I IFN expression. Tofacitinib also decreased the T cell stimulatory capability of DCs and increased expression of indoleamine 2,3-dioxygenase (IDO)-1 and IDO-2. Tofacitinib, a JAK1/JAK3 inhibitor, affected the activities of human DCs. It decreased CD80/CD86 expression and T cell stimulatory capability through suppression of type I IFN signalling. These results suggest a novel mode of action for tofacitinib and a pivotal role for JAKs in the differentiation of DCs.
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Supelco
Pyrrole, analytical standard
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, for fluorescence, free acid