Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Neodymium, rod, 50mm, diameter 6.35mm, cast, 99%
SKU
Pack Size
Availability
Price
Quantity
Neodymium, rod, 100mm, diameter 12.5mm, cast, 99%
SKU
Pack Size
Availability
Price
Quantity
Neodymium, rod, 100mm, diameter 6.35mm, cast, 99%
SKU
Pack Size
Availability
Price
Quantity
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Praseodymium(III) nitrate hexahydrate, 99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
SKU
Pack Size
Availability
Price
Quantity
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
USP
Citric acid, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, 99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, puriss. p.a., ≥98.5% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Nickel(II) nitrate hexahydrate, purum p.a., crystallized, ≥97.0% (KT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade