Merck
  • Presence of disinfectant resistance genes in Escherichia coli isolated from retail meats in the USA.
Presence of disinfectant resistance genes in Escherichia coli isolated from retail meats in the USA.
The Journal of antimicrobial chemotherapy (2014-06-09)
Likou Zou, Jianghong Meng, Patrick F McDermott, Fei Wang, Qianru Yang, Guojie Cao, Maria Hoffmann, Shaohua Zhao
ABSTRACT
To examine the distribution of all genes known to be responsible for resistance to quaternary ammonium compounds (QACs), and their association with resistance to QACs and other antimicrobials, in Escherichia coli recovered from retail meats. A total of 570 strains of E. coli isolated from US retail meats in 2006 were screened for the presence of 10 QAC resistance genes [qacE, qacEΔ1, qacF, qacG, emrE, sugE(c), sugE(p), mdfA and ydgE/ydgF]. The MICs of six common disinfectants were determined using an agar dilution method. Possible associations between the presence of the gene and bacterial resistance to QACs and antimicrobials were investigated. emrE, sugE(c), mdfA and ydgE/ydgF were commonly present (77.2%-100%) in the E. coli isolates, but qac and sugE(p) were less prevalent (0.4%-22.3%). emrE-mdfA-sugE(c)-ydgE/F was the most common QAC resistance gene profile. A significant association was found between antimicrobial resistance and the presence of sugE(p) and qacEΔ1 (P < 0.05). Antimicrobial-resistant E. coli isolates tended to contain more diverse combinations of disinfectant resistance genes than susceptible ones. All isolates showed reduced susceptibility to five of six disinfectants compared with the control strains. Higher MICs were generally associated with the presence of qac and sugE(p) genes. The QAC resistance genes were commonly present among E. coli isolated from retail meats, and the qac and sugE(p) genes were highly associated with multidrug resistance phenotypes. Using QACs in the food industry may not be as effective as expected and could provide selection pressure for strains with acquired resistance to other antimicrobials.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Chlorhexidine, ≥99.5%
Chlorhexidine, European Pharmacopoeia (EP) Reference Standard
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Triclosan, United States Pharmacopeia (USP) Reference Standard
Supelco
Chlorhexidine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard