Merck
  • Isoniazid conjugated poly(lactide-co-glycolide): long-term controlled drug release and tissue regeneration for bone tuberculosis therapy.
Isoniazid conjugated poly(lactide-co-glycolide): long-term controlled drug release and tissue regeneration for bone tuberculosis therapy.
Biomaterials (2015-03-31)
Da Huang, Dawei Li, Tiantian Wang, Hong Shen, Pei Zhao, Baoxia Liu, Yezi You, Yuanzheng Ma, Fei Yang, Decheng Wu, Shenguo Wang
ABSTRACT
Bone tuberculosis (TB) is one of the most common extrapulmonary TB. Effective integration of chemotherapy and bone regeneration is an optimal solution for bone TB therapy. Herein, we produce a composite scaffold drug delivery system fabricated with an isoniazid conjugated star poly(lactide-co-glycolide) (PLGA-INH4) and β-TCP. The cytological assay indicated the composite system possesses good biocompatibility. The in vitro and in vivo drug release evaluations showed that the composite system can intactly release the pristine INH and maintain effective INH concentration in a controlled manner for more than 100 days, and achieve high localized drug concentration and low systemic drug concentration. The rabbit radius repair experiment testified the scaffold has good bone regeneration capacity. Our work demonstrate the composite system can simultaneously achieve localized long-term drug controlled release and bone regeneration, which provides a promising route for improved bone TB treatment.
Product Description
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Supelco
Isoniazid, Vetec, reagent grade, 98%
Tantalum(V) ethoxide, packaged for use in deposition systems
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Isoniazid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Supelco
1,4-Dioxane, analytical standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Pentaerythritol, 98%
Sigma-Aldrich
1,4-Dioxane, anhydrous, 99.8%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
4-Formylbenzoic acid, 97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
1,4-Dioxane, ReagentPlus®, ≥99%, contains ≤25 ppm BHT as stabilizer