Merck
  • Isolation and drug susceptibility of Candida parapsilosis sensu lato and other species of C. parapsilosis complex from patients with blood stream infections and proposal of a novel LAMP identification method for the species.
Isolation and drug susceptibility of Candida parapsilosis sensu lato and other species of C. parapsilosis complex from patients with blood stream infections and proposal of a novel LAMP identification method for the species.
Mycopathologia (2014-12-08)
Plinio Trabasso, Tetsuhiro Matsuzawa, Renata Fagnani, Yasunori Muraosa, Kenichiro Tominaga, Mariangela Ribeiro Resende, Katsuhiko Kamei, Yuzuru Mikami, Angelica Zaninelli Schreiber, Maria Luiza Moretti
ABSTRACT
Candida parapsilosis complex (CPC) is the third Candida species isolated in blood cultures of patients from our Hospital, following C. albicans and C. tropicalis. From 2006 to 2010, the median annual distribution of CPC was 8 cases/year. Records of 36 patients were reviewed. CPC were 31 (86.1%) C. parapsilosis; 4 (11.1%) C. orthopsilosis; and 1 (2.8%) C. metapsilosis. Clinical characteristics were central venous catheter, 34 (94.4%); parental nutrition, 25 (70%); surgery, 27 (57.9%); prior bacteremia, 20 (51.3%); malignancy, 18 (50%). General mortality was 47.2%. Death was higher in immunosuppressed patients (17 vs. 11; p = 0.003). Three out four (75%) patients with C. orthopsilosis and 14 out 31 (45.2%) with C. parapsilosis died (p = 0.558). Thirty-nine individual isolates were tested for susceptibility to seven antifungal drugs, with MICs values showing susceptibility to all of them. Two isolates, one C. orthopsilosis and one C. parapsilosis, had fluconazole MIC = 4 μg/mL. Differentiation among CPC has implication in caring for patients with invasive candidiasis since there are differences in virulence, pathogenicity and drug susceptibility. A method targeting the topoisomerase II gene based on loop-mediated isothermal amplification (LAMP) was developed. LAMP emerges as a promising tool for the identification of fungal species due to the high sensitivity and specificity. LAMP can be performed at the point-of-care, being no necessary the use of expensive equipment. In our study, the method was successful comparing to the DNA sequencing and proved to be a reliable and fast assay to distinguish the three species of CPC.
Product Description
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Voriconazole, VETRANAL®, analytical standard
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Voriconazole, European Pharmacopoeia (EP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Flucytosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
5-Fluorocytosine, nucleoside analog
Itraconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Flucytosine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Miconazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Miconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%