Merck
  • Role of seaweed laminaran from Saccharina longicruris on matrix deposition during dermal tissue-engineered production.
Role of seaweed laminaran from Saccharina longicruris on matrix deposition during dermal tissue-engineered production.
International journal of biological macromolecules (2015-01-21)
Akram Ayoub, Jadson Moreira Pereira, Laurie-Eve Rioux, Sylvie L Turgeon, Martin Beaulieu, Véronique J Moulin
ABSTRACT
Our laboratory has developed a technique to reconstruct in vitro tissue from human cells using the self-assembly tissue-engineering method, which utilizes the ability of fibroblasts to deposit the matrix they secrete. The time necessary for tissue construction, several weeks, is a drawback for many clinical uses. We hypothesized that the addition of laminaran can increase the deposition of matrix, speeding up the production of the tissue. Laminaran was isolated from the brown seaweed Saccharina longicruris harvested in Canada and its structure was evaluated. Laminaran is a small molecular weight polysaccharide composed of linear glucose chains. Monolayer-cultured human skin fibroblasts were cultured in the presence of laminaran with ascorbate for 7 or 35 days to produce a dermis. Treatment did not induce any variation in the growth rate or alpha smooth muscle actin content but it did increase the deposition of collagen I in a dose-dependent manner. After 35 days, the reconstructed dermal thickness was increased when laminaran was added, and collagen I deposition and MMP activity were also significantly increased. Thus, laminaran can be used to increase the rate of production of reconstructed self-assembled dermis and can also potentially be used in cosmetic or therapeutic creams to stimulate matrix production.
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane