Merck
Dysbindin-1, a schizophrenia-related protein, interacts with HDAC3.
Neuroscience letters (2014-09-10)
Mika Soma, Min Wang, Satoshi Suo, Shoichi Ishiura
ABSTRACT
DTNBP1 is a key candidate gene associated with schizophrenia. The expression of its protein product, dysbindin-1, is altered in the brains of schizophrenic patients; however, the physiological functions of dysbindin-1 in the central nervous system are unclear. Several studies have shown that both dysbindin-1 and histone deacetylase 3 (HDAC3) can be phosphorylated by the DNA-dependent protein kinase complex. In this study, we investigated the relationship between dysbindin-1 and HDAC3. We found that dysbindin-1 formed a protein complex with HDAC3 in human neuroblastoma cells and in mouse brain. The interaction between dysbindin-1 and HDAC3 occurred in an isoform-specific manner: HDAC3 coupled with dysbindin-1A and -1B, but not -1C. We also found that dysbindin-1B expression was increased in the nucleus in the presence of HDAC3, and, conversely, that the phosphorylation level of HDAC3 increased in the presence of dysbindin-1B. Taken together, these results identify a novel binding partner for dysbindin-1, which may potentially provide a new avenue for research into the neurological mechanisms of schizophrenia.
Sigma-Aldrich
Anti-phospho-HDAC3 (pSer424) antibody produced in rabbit, affinity isolated antibody