Merck
  • eHealth: individualization of mesalazine treatment through a self-managed web-based solution in mild-to-moderate ulcerative colitis.
eHealth: individualization of mesalazine treatment through a self-managed web-based solution in mild-to-moderate ulcerative colitis.
Inflammatory bowel diseases (2014-09-24)
Natalia Pedersen, Peter Thielsen, Lars Martinsen, Mette Bennedsen, Anne Haaber, Ebbe Langholz, Zsuzsanna Végh, Dana Duricova, Tine Jess, Sally Bell, Johan Burisch, Pia Munkholm
ABSTRACT
To individualize treatment with mesalazine for ulcerative colitis relapses through a self-managed, web-based solution to optimize the short-term disease course. Prospective, open-label, web-guided study with 3 months mesalazine therapy among patients with mild-to-moderate ulcerative colitis. Once a week, patients completed the simple clinical colitis activity index (SCCAI) and registered fecal calprotectin (FC) on the web application: www.meza.constant-care.dk. SCCAI and FC were summed and resulted in a total inflammatory burden score (TIBS). Deep remission was defined as SCCAI ≤1; FC = 0, and TIBS ≤1. A total of 95 patients (62% females; median age 45 yr) were included in the study and allocated 4.8 g mesalazine per day. Of these, 82 (86%) patients were adherent to web therapy, completing 3 months of web-guided mesalazine therapy. Of the 82 adherent patients, 72 (88%) continued mesalazine and 10 (12%) needed rescue therapy. From weeks 0 to 12, patients had experienced a significant reduction in mean SCCAI (4.6 versus 1.6, P < 0.001), mean FC (437 versus 195, P < 0.001), and mean TIBS (6.7 versus 2.4, P < 0.001). Based on TIBS values (≤1), the dose of mesalazine was reduced to 2.4 g in 25% of patients at week 3 in 50% of subjects at week 5 and in 88% of patients at week 12. Web-guided therapy with mesalazine in mild-to-moderate ulcerative colitis helps to individualize the dose and improve adherence to therapy. The study confirms mesalazine efficacy in mild-to-moderate UC, significantly improving TIBS values in majority of the patients.
Product Description
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Aminosalicylic acid, 95%
Sigma-Aldrich
5-Aminosalicylic acid, ≥99%
Supelco
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
Mesalazine, European Pharmacopoeia (EP) Reference Standard