Merck
  • Attenuation of allergic immune response phenotype by mannosylated egg white in orally induced allergy in BALB/c mice.
Attenuation of allergic immune response phenotype by mannosylated egg white in orally induced allergy in BALB/c mice.
Journal of agricultural and food chemistry (2014-09-13)
Prithy Rupa, Soichiro Nakamura, Shigeru Katayama, Yoshinori Mine
ABSTRACT
Food allergies are attributed to an imbalance in immune response to ubiquitous antigens. A previous study demonstrated that mannose glycation (mannosylation) of ovalbumin decreased allergenicity in vivo. The proposed research targets mannosylation of various common allergens that may help prevent food allergy. Balb/c mice (n = 8) were sensitized toxin egg white, peanut, and whey and treated with mannosylated forms of the test antigens. Glucosylated peanut and cholera toxins were used as controls. Allergic status was assessed as clinical signs, serum histamine, mouse mast cell protease (MMCP), antibody activity, cytokines, and T regulatory cells (T-regs). Significant preventative effects were observed with mannosylated egg white treatment such as reduced clinical signs, histamine, MMCP, specific G, G1, and E antibody activities, and IL-4 and increased IL-10 and CD25(+) Foxp3(+) cells. Other groups did not differ significantly. It was concluded that mannosylated egg white provides a powerful tool to prevent allergic phenotypes with possible relevance to control human egg allergy.
Product Description
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Supelco
Histamine, analytical standard
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)
Sigma-Aldrich
D-(+)-Glucose, Vetec, reagent grade, ≥99.5% (HPLC)