Merck
  • Toll-like receptor-mediated responses by placental Hofbauer cells (HBCs): a potential pro-inflammatory role for fetal M2 macrophages.
Toll-like receptor-mediated responses by placental Hofbauer cells (HBCs): a potential pro-inflammatory role for fetal M2 macrophages.
American journal of reproductive immunology (New York, N.Y. : 1989) (2014-10-28)
Omar M Young, Zhonghua Tang, Tracy Niven-Fairchild, Serkalem Tadesse, Graciela Krikun, Errol R Norwitz, Gil Mor, Vikki M Abrahams, Seth Guller
ABSTRACT
Microbial-driven responses in placenta are linked with adverse pregnancy outcomes. The role of Toll-like receptor (TLR) function in Hofbauer cells (HBCs) and fetal macrophages of the placental villous core remains understudied. Flow cytometry and immunohistochemistry (IHC) were used to establish the phenotype of HBCs. Regulation of cytokine secretion in response to treatment with TLR agonists and expression levels of TLRs and co-activators were compared in HBCs, placental fibroblasts (FIBs), and human umbilical vein endothelial cells (HUVECs) using ELISA and qPCR. Although flow cytometry and IHC revealed HBCs to be M2 (anti-inflammatory) macrophages, LPS and polyinosinic: polycytidylic acid [poly (I:C)] treatments markedly enhanced IL-6 secretion by HBCs, and expression of TLR-2, TLR-3, TLR-4, CD14, and MD-2 was the highest in HBCs. These results indicate that although HBCs are M2 macrophages, inflammatory responses are induced through TLR-3 and TLR-4 in this cell type, suggesting a role in microbial-driven placental/fetal inflammation.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Linoleic acid, analytical standard
SAFC
HEPES
Sigma-Aldrich
L-Ascorbic acid, 99%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Methanol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
SAFC
HEPES
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)