Merck
  • Effervescent-salt-assisted dispersive micro-solid-phase extraction using mesoporous hybrid materials coupled with ultra-performance liquid chromatography for the determination of trace-level compounds in complicated plant preparations.
Effervescent-salt-assisted dispersive micro-solid-phase extraction using mesoporous hybrid materials coupled with ultra-performance liquid chromatography for the determination of trace-level compounds in complicated plant preparations.
Analytical and bioanalytical chemistry (2014-12-21)
Li-Hong Ye, Wan Cao, Shuai-Shuai Hu, Jian-Hua Da, Han-Bin Dai, Jun Cao, Jing-Jing Xu, Xiao-Qing Pang
ABSTRACT
A novel effervescent-salt-assisted dispersive micro-solid-phase extraction using mesoporous hybrid materials was developed for the extraction of minute traces of constituents in complicated plant preparations. In this study, a special tablet containing carbon dioxide sources (sodium dihydrogenphosphate and sodium carbonate) and the sorbent (mesoporous hybrid materials) was prepared. The effects of different parameters influencing the extraction efficiency such as the concentration of salts, the type and concentration of mesoporous material, pH of diluent, and desorption solvents were investigated and optimized. Results show that the proposed method using green solvents (water) as extraction solutions required low consumption of sample amount and obtained high enrichment efficiency. Moreover, under optimized conditions, the tested tanshinones exhibited good linearity (r(2) > 0.994) in the concentration range of 0.5 to 80 ng mL(-1). The limits of detection values were lower than 0.0803 pg using UV-visible detection. The developed method was successfully applied for the analysis of trace tanshinones in compound Danshen dripping pill and Danqi tablet samples.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis