Merck
  • Determination of fluoroquinolones in chicken feces - a new liquid-liquid extraction method combined with LC-MS/MS.
Determination of fluoroquinolones in chicken feces - a new liquid-liquid extraction method combined with LC-MS/MS.
Environmental toxicology and pharmacology (2014-10-13)
Franziska Janusch, Gesine Scherz, Siegrun A I Mohring, Gerd Hamscher
ABSTRACT
The application of antibiotics including fluoroquinolones to farming animals is widespread and may lead to the development of antibiotic resistance and other environmental effects. To calculate environmental loads and for a proper risk assessment it is necessary to determine the antibiotic concentration in feces. Therefore, a new liquid-liquid extraction method combined with HPLC-MS/MS for the detection of marbofloxacin, ciprofloxacin, enrofloxacin and difloxacin in chicken feces was developed. Recoveries ranged from 51.0% to 83.5%. LOQs were between 0.10 and 1.09μg/kg. Feces of chickens treated with an enrofloxacin dosage of 10mg/kg bodyweight revealed maximum enrofloxacin and ciprofloxacin concentrations of 61.3 and 18.8mg/kg. Both antibiotics could be detected in feces up to two days after the last application in notable amounts (∼1mg/kg). Thus, feces of recently medicated chickens should not be used as a fertilizer without any further processing.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonia, puriss., anhydrous, ≥99.95%
Sigma-Aldrich
Diethyl ether, puriss. p.a., ACS reagent, ≥99.8% (GC)
Supelco
Acetone, analytical standard
Supelco
tert-Butyl methyl ether, suitable for HPLC, ≥99.8% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Marbofloxacin, VETRANAL®, analytical standard
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Marbofloxacin, European Pharmacopoeia (EP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%