Merck
  • Zebrafish liver (ZFL) cells are able to mount an anti-viral response after stimulation with Poly (I:C).
Zebrafish liver (ZFL) cells are able to mount an anti-viral response after stimulation with Poly (I:C).
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2014-12-24)
Angels Ruyra, Debora Torrealba, Davinia Morera, Lluís Tort, Simon MacKenzie, Nerea Roher
ABSTRACT
The zebrafish (Danio rerio) is a widely used model species for biomedical research and is also starting to be a model for aquaculture research. The ZFL cell line, established from zebrafish liver, has been mostly used in toxicological and ecotoxicological studies. However, no studies have previously characterised this cell line in regard to its immunological response. The aim of this work was to study the gene expression response of the ZFL cell line after incubation with different prototypical immune stimuli, such as lipopolysaccharide (LPS), peptidoglycan (PGN), zymosan, and with a special focus on the dsRNA Poly (I:C). Using PCR, microarrays, and confocal microscopy we have explored the response of the ZFL cells against Poly (I:C). This study shows that the ZFL is able to uptake very efficiently the Poly (I:C) and mount a strong anti-viral response. We can conclude that ZFL could be used not only in toxicological studies, but also in studying anti-viral responses in zebrafish.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%