Merck
  • Immunoglobulin M oligoclonal bands: biomarker of targetable inflammation in primary progressive multiple sclerosis.
Immunoglobulin M oligoclonal bands: biomarker of targetable inflammation in primary progressive multiple sclerosis.
Annals of neurology (2014-06-10)
Luisa M Villar, Bonaventura Casanova, Nadia Ouamara, Manuel Comabella, Farzaneh Jalili, David Leppert, Clara de Andrés, Guillermo Izquierdo, Rafael Arroyo, Timuçin Avşar, Sergey V Lapin, Trina Johnson, Xavier Montalbán, Oscar Fernández, Roberto Álvarez-Lafuente, Donna Masterman, María I García-Sánchez, Francisco Coret, Aksel Siva, Evgeniy Evdoshenko, José C Álvarez-Cermeño, Amit Bar-Or
ABSTRACT
To identify a biomarker distinguishing patients who, despite a primary progressive multiple sclerosis (PPMS) clinical course, may nonetheless benefit from immune therapy. The presence or absence of both immunoglobulin (Ig) G and IgM oligoclonal bands (OCB) was blindly examined in paired cerebrospinal fluid (CSF) and serum samples from a large PPMS patient cohort, and related to clinical and imaging evidence of focal inflammatory disease activity. Using both cross-sectional samples and serial sampling in a subgroup of patients followed prospectively as part of the placebo-controlled OLYMPUS study of rituximab in PPMS, we found that the presence of CSF-restricted IgM OCB (but not of IgG OCB) is associated with an active inflammatory disease phenotype in PPMS patients. This finding was confirmed in an independent, multicenter validation cohort. The presence of CSF IgM OCB may be a biomarker for a subset of PPMS patients with more active inflammatory disease, who may benefit from immune-directed treatments.
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)