Merck
  • Analgesic effects of clinically used compounds in novel mouse models of polyneuropathy induced by oxaliplatin and cisplatin.
Analgesic effects of clinically used compounds in novel mouse models of polyneuropathy induced by oxaliplatin and cisplatin.
Neuro-oncology (2014-04-10)
Jennifer R Deuis, Yu Ling Lim, Silmara Rodrigues de Sousa, Richard J Lewis, Paul F Alewood, Peter J Cabot, Irina Vetter
ABSTRACT
Peripheral neuropathy is the major dose-limiting side effect of cisplatin and oxaliplatin, and there are currently no effective treatments available. The aim of this study was to assess the pharmacological mechanisms underlying chemotherapy-induced neuropathy in novel animal models based on intraplantar administration of cisplatin and oxaliplatin and to systematically evaluate the analgesic efficacy of a range of therapeutics. Neuropathy was induced by a single intraplantar injection of cisplatin or oxaliplatin in C57BL/6J mice and assessed by quantification of mechanical and thermal allodynia. The pharmacological basis of cisplatin-induced neuropathy was characterized using a range of selective pharmacological inhibitors. The analgesic effects of phenytoin, amitriptyline, oxcarbazepine, mexiletine, topiramate, retigabine, gabapentin, fentanyl, and Ca(2+/)Mg(2+) were assessed 24 hours after induction of neuropathy. Intraplantar administration of cisplatin led to the development of mechanical allodynia, mediated through Nav1.6-expressing sensory neurons. Unlike intraplantar injection of oxaliplatin, cold allodynia was not observed with cisplatin, consistent with clinical observations. Surprisingly, only fentanyl was effective at alleviating cisplatin-induced mechanical allodynia despite a lack of efficacy in oxaliplatin-induced cold allodynia. Conversely, lamotrigine, phenytoin, retigabine, and gabapentin were effective at reversing oxaliplatin-induced cold allodynia but had no effect on cisplatin-induced mechanical allodynia. Oxcarbazepine, amitriptyline, mexiletine, and topiramate lacked efficacy in both models of acute chemotherapy-induced neuropathy. This study established a novel animal model of cisplatin-induced mechanical allodynia consistent with the A-fiber neuropathy seen clinically. Systematic assessment of a range of therapeutics identified several candidates that warrant further clinical investigation.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Oxaliplatin, powder
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard