Merck
  • Recovery of redox homeostasis altered by CuNPs in H4IIE liver cells does not reduce the cytotoxic effects of these NPs: an investigation using aryl hydrocarbon receptor (AhR) dependent antioxidant activity.
Recovery of redox homeostasis altered by CuNPs in H4IIE liver cells does not reduce the cytotoxic effects of these NPs: an investigation using aryl hydrocarbon receptor (AhR) dependent antioxidant activity.
Chemico-biological interactions (2015-01-27)
Mona Connolly, María Luisa Fernández-Cruz, José María Navas
ABSTRACT
The generation of reactive oxygen species (ROS) and consequent oxidative stress is regarded as a relevant mechanism for nanoparticle toxicity. In cells, the activation of the aryl hydrocarbon receptor (AhR) triggers a cascade of defence responses against oxidative stress. By increasing AhR dependent cellular anti-oxidant activity, we tested the extent to which the cytotoxic effect of copper nanoparticles (CuNPs) is governed by oxidative stress. H4IIE rat hepatoma cells were challenged with high ROS levels after exposure to CuNPs, while the AhR-induced cellular anti-oxidant defence was simultaneously activated by the AhR ligand beta-Naphthoflavone (ßNF). Activation of phase II detoxification enzymes (as glutathione-S-transferases, GSTs) and anti-oxidants (glutathione, GSH) led to a complete abrogation of CuNP-induced ROS production. However, a concurrent reduction in cytotoxicity was not detected, thereby indicating that CuNPs exert non-oxidative stress mediated cytotoxic effects. Transmission electron microscopy analysis pointed to a direct physical perturbation of cellular structures by CuNPs, thus contributing to their cytotoxicity. Our observations highlight that distinct mechanisms underlie the toxicity of ions and NPs and indicate that while ROS elicitation is CuNP-specific, the cytotoxic action of these particles may not be directly related to their pro-oxidative activity. These findings have important implications with respect to the oxidative stress paradigm used to explain NP toxicity.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
1-Chloro-2,4-dinitrobenzene, 97%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), Vetec, reagent grade, 98%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
L-Glutathione oxidized disodium salt, ≥98%, powder
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
L-Glutathione oxidized disodium salt, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Fluorescamine, ≥98% (TLC), powder, used for detection of primary amines
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, Vetec, reagent grade, ≥98%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder