Merck
  • Quantitative multiplexed detection of common pulmonary fungal pathogens by labeled primer polymerase chain reaction.
Quantitative multiplexed detection of common pulmonary fungal pathogens by labeled primer polymerase chain reaction.
Archives of pathology & laboratory medicine (2014-10-31)
Zhengming Gu, Daelynn R Buelow, Ruta Petraitiene, Vidmantas Petraitis, Thomas J Walsh, Randall T Hayden
ABSTRACT
Invasive fungal infections are an important cause of morbidity and mortality among immunocompromised patients. To design and evaluate a multiplexed assay aimed at quantitative detection and differentiation of the 5 molds that are most commonly responsible for pulmonary infections. Using labeled primer polymerase chain reaction chemistry, an assay was designed to target the 5.8S and 28S ribosomal RNA genes of Aspergillus spp, Fusarium spp, Scedosporium spp, and members of the order Mucorales ( Rhizopus oryzae , Rhizopus microsporus, Cunninghamella bertholletiae, Mucor circinelloides, Lichtheimia corymbifera, and Rhizomucor pusillus). This assay was split into 2 multiplexed reactions and was evaluated using both samples seeded with purified nucleic acid from 42 well-characterized clinical fungal isolates and 105 archived samples (47 blood [45%], 42 bronchoalveolar lavage fluid [40%], and 16 tissue [15%]) collected from rabbit models of invasive pulmonary fungal infections. Assay detection sensitivity was less than 25 copies of the target sequence per reaction for Aspergillus spp, 5 copies for Fusarium spp and Scedosporium spp, and 10 copies for the Mucorales. The assay showed quantitative linearity from 5 × 10(1) to 5 × 10(5) copies of target sequence per reaction. Sensitivities and specificities for bronchoalveolar lavage fluid, tissue, and blood samples were 0.86 and 0.99, 0.60 and 1.00, and 0.46 and 1.00, respectively. Labeled primer polymerase chain reaction permits rapid, quantitative detection and differentiation of common agents of invasive fungal infection. The assay described herein shows promise for clinical implementation that may have a significant effect on the rapid diagnosis and treatment of patients' severe infections caused by these pulmonary fungal pathogens.
Product Description
Sorbitol, European Pharmacopoeia (EP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Ceftazidime, European Pharmacopoeia (EP) Reference Standard
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Cytarabine, European Pharmacopoeia (EP) Reference Standard
USP
Cytarabine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sodium phosphate monobasic, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
6α-Methylprednisolone, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%