Merck
  • Cytochrome P450 3A-mediated metabolism of the topoisomerase I inhibitor 9-aminocamptothecin: impact on cancer therapy.
Cytochrome P450 3A-mediated metabolism of the topoisomerase I inhibitor 9-aminocamptothecin: impact on cancer therapy.
International journal of oncology (2014-06-04)
Alexandra Maier-Salamon, Theresia Thalhammer, Gottfried Reznicek, Michaela Böhmdorfer, István Zupkó, Alexander Hartl, Walter Jaeger
ABSTRACT
The metabolism of 9-aminocamptothecin (9-AC) was investigated in human and rat liver microsomes. In both species 9-AC was almost exclusively biotransformed to dihydroxy-9-AC (M1) and monohydroxy-9-AC (M2). The enzymatic efficiencies of the formation of M1 and M2 (V(max)/K(m)) were 1.7- and 2.7‑fold higher in rat than in human liver microsomes indicating species-related differences in 9-AC hydroxylation. Incubation in the presence of human recombinant cytochrome P450 (CYP) enzymes demonstrated that the formation of M1 and M2 is mainly catalyzed by CYP3A4 and only to a minor extent by extrahepatic CYP1A1. The predominant role of CYP3A4 was further supported by a dramatic inhibition of metabolite formation in the presence of the CYP3A4 substrates troleandomycin and ketoconazole. Experiments conducted in isolated perfused rat livers further demonstrated that biliary excretion of 9-AC, M1 and M2 during 60 min of perfusion was pronounced and accounted for 17.7±2.59, 0.05±0.01 and 2.75±0.14% of total 9-AC applied to the liver, respectively. In summary, this study established that CYP3A-dependent hydroxylation is the main metabolic pathway for 9-AC in rat and human liver, which have to be taken into consideration during cancer therapy of patients.
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid trisodium salt, 98-100%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Ketoconazole, United States Pharmacopeia (USP) Reference Standard
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Methanol, analytical standard