Merck
  • Umbilical cord-derived mesenchymal stem cells instruct dendritic cells to acquire tolerogenic phenotypes through the IL-6-mediated upregulation of SOCS1.
Umbilical cord-derived mesenchymal stem cells instruct dendritic cells to acquire tolerogenic phenotypes through the IL-6-mediated upregulation of SOCS1.
Stem cells and development (2014-04-16)
Yinan Deng, Shuhong Yi, Guoying Wang, Jintao Cheng, Yingcai Zhang, Wenjie Chen, Yan Tai, Shaohong Chen, Guihua Chen, Wei Liu, Qi Zhang, Yang Yang
ABSTRACT
The mechanisms responsible for the inhibitory effects of mesenchymal stem cells (MSCs) on dendritic cells (DCs) are still poorly understood. Our investigation of the potential signaling pathways revealed for the first time that human umbilical-cord-derived MSCs (UC-MSCs) instruct DCs to acquire tolerogenic phenotypes through the IL-6-mediated upregulation of SOCS1. This subset of MSC-DCs exhibited a tolerogenic pattern, with a clear decrease in the expression of co-stimulatory molecules and the capacity to stimulate CD3(+) T cell proliferation and inflammatory factor secretion, and a significant increase in the production of inhibitory cytokine IL-10 and the ability to induce Treg cells and Th2 responses. Adoption of this tolerogenic pattern required the activation of SOCS1, which blocked DC maturation by impairing TLR4 signaling. The effects of UC-MSCs on SOCS1 activation were essentially mediated by the JAK-STAT pathway via IL-6 secretion. In summary, our data identify a new mechanism, involving the IL-6-mediated upregulation of SOCS1, by which UC-MSCs instruct DCs to acquire tolerogenic phenotypes.
Sigma-Aldrich
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat IL-10 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine