Merck
  • Improvement of the diagnostic accuracy of MRA with subtraction technique in cerebral vasospasm.
Improvement of the diagnostic accuracy of MRA with subtraction technique in cerebral vasospasm.
Journal of neuroimaging : official journal of the American Society of Neuroimaging (2014-07-22)
Akiyoshi Hamaguchi, Noriyuki Fujima, Daisuke Yoshida, Naoko Hamaguchi, Shuichi Kodera
ABSTRACT
Vasospasm has been considered the most severe acute complication after subarachnoid hemorrhage (SAH). MRA is not considered ideal for detecting cerebral vasospasm because of background including the hemorrhage. The aim of this study is to evaluate the efficacy of Subtraction MRA (SMRA) by comparing it to that of conventional MRA (CMRA) for diagnosis of cerebral vasospasm. Arteries were assigned to one of three categories based on the degree of MRA diagnostic quality of vasospasm (quality score): 0, bad … 2, good. Furthermore each artery was assigned to one of four categories based on the degree of vasospasm severity (SV score): 0, no vasospasm … 3, severe. The value of the difference between DSA-SV score and MRA-SV score was defined as the DIF score. CMRA and SMRA were compared for each arterial region with regard to quality score and DIF score. The average CMRA and SMRA quality score were 1.46 and 1.79; the difference was statistically significant. The average CMRA and SMRA DIF score were 1.08 and .60; the difference was statistically significant. Diagnosis of cerebral vasospasm is more accurate by SMRA than by CMRA. The advantages are its noninvasive nature and its ability to detect cerebral vasospasm.
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)