Merck
  • Alternative QuEChERS-based modular approach for pesticide residue analysis in food of animal origin.
Alternative QuEChERS-based modular approach for pesticide residue analysis in food of animal origin.
Analytical and bioanalytical chemistry (2015-03-17)
Karolina Lichtmannegger, Roman Fischer, Franz Xaver Steemann, Hermann Unterluggauer, Sonja Masselter
ABSTRACT
The approach for pesticide residue analysis in food of animal origin differs strongly from the one established for food of plant origin, as laboratories mainly focus on conventional methods for the analysis of non-polar pesticides known to accumulate in fatty tissues. However, these group-specific methods are very laborious and cost intensive and typically require extraction of fat components followed by extensive clean-up steps to remove matrix constituents. This work highlights the development and validation of a straightforward QuEChERS-derived clean-up procedure enabling facile, precise, and reliable identification and quantitation of pesticide residues in food of animal origin, which can be extended to various other commodities with moderate fat content and applied to replace traditional group-specific methods. Two additional methods for lean and highly fatty commodities complete this well-established "simplified modular system". The proposed method was in-house validated in terms of accuracy and precision, recovery and linearity as well as specificity and sensitivity on two representative commodities for food of animal origin (egg and meat). For the majority of the more than 80 pesticides investigated, satisfactory results were obtained. Procedural matrix calibration was applied for screening purposes in conjunction with GC-MS/MS or LC-MS/MS as integral part of the approach. Apparent recoveries for most of the compounds ranged from 70 to 120 % (RSD < 20%) at spiking levels of 0.01 (LOQ level) and 0.05 mg/kg. Accuracy of the method has been demonstrated by application to reference material from previous EU proficiency tests. Only in case of positive screening results a standard addition approach was applied for precise quantitation.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG