Merck
  • The effects of lobeline on nicotine withdrawal-induced depression-like behavior in mice.
The effects of lobeline on nicotine withdrawal-induced depression-like behavior in mice.
Psychopharmacology (2014-04-01)
Monzurul Amin Roni, Shafiqur Rahman
ABSTRACT
Evidence suggests that neuronal nicotinic acetylcholine receptor (nAChR) ligand lobeline has antidepressant-like properties. The present study investigated the effects of lobeline on nicotine withdrawal-induced depression-like behavior. Adult C57BL/6J mice were exposed to nicotine (200 μg/ml) in drinking solution for 3 weeks. During withdrawal, depression-like behavior was measured by the forced swim test (FST). We also determined norepinephrine (NE) levels in the prefrontal cortex (PFC) and hippocampus during nicotine withdrawal. Furthermore, we determined the effects of repeated treatment with lobeline or a selective α4β2 nAChR ligand 3-(pyridine-3́-yl)-cytisine on brain-derived neurotrophic factor (BDNF) and phosphorylated cAMP-responsive element binding (p-CREB) protein expression in the hippocampus. Withdrawal from chronic nicotine increased immobility time in the FST, a measure for depression-like behavior. Pretreatment with lobeline significantly decreased immobility time during nicotine withdrawal. In addition, pretreatment with lobeline attenuated nicotine withdrawal-induced increased NE levels in the PFC and hippocampus. Further, repeated treatment with lobeline or 3-(pyridine-3́-yl)-cytisine decreased immobility time in the FST and reduced withdrawal-induced increased BDNF and p-CREB expression in the hippocampus. Taken together, our results indicate that lobeline attenuated nicotine withdrawal-induced depression-like behavior likely by targeting brain nAChRs, noradrenergic neurotransmission, and/or hippocampal BDNF. Thus, lobeline may have some potential to prevent smoking relapse by counteracting nicotine withdrawal-induced depression in humans.
Product Description
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
Bupropion hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
USP
Bupropion hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(−)-Lobeline hydrochloride, 98%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Lobeline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)