Merck
  • Rapid analysis of trace volatile formaldehyde in aquatic products by derivatization reaction-based surface enhanced Raman spectroscopy.
Rapid analysis of trace volatile formaldehyde in aquatic products by derivatization reaction-based surface enhanced Raman spectroscopy.
The Analyst (2014-05-31)
Zhuomin Zhang, Cheng Zhao, Yunjian Ma, Gongke Li
ABSTRACT
Toxic formaldehyde is sometimes used illegally as a food preservative, however, on-site rapid analysis of trace formaldehyde in aquatic products remains a challenge. In this work, a simple on-site rapid quantification method for trace volatile formaldehyde in aquatic products was developed by a derivative reaction-based surface enhanced Raman spectroscopy (SERS) technique coupled with a homemade portable purge-sampling device. Trace formaldehyde separated from complicated aquatic matrices via a purge-sampling procedure was reacted with a derivative reagent to produce a Raman-active analyte for consequent SERS analysis. Au/SiO2 nanoparticles (NPs) were employed as the enhancement substrate to achieve significant enhancement of Raman signal intensity. Conditions of derivative reaction and SERS detection were optimized in detail, and the selectivity of this analytical method was also evaluated based on related analogs. Under optimal conditions, an extremely low detection limit of 0.17 μg L(-1) was achieved. Trace volatile formaldehyde can be found in fresh squid and shrimp samples without obvious matrix interference, and this was quantified to be 0.13-0.21 mg kg(-1) using the described method. The recoveries of spiked aquatic product samples were found to be 70.0-89.1% with RSDs of 2.3-7.2% (n = 3). The results suggest that the proposed method is reliable and suitable for on-site rapid analysis of trace formaldehyde in aquatic products.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Butyraldehyde, ≥96.0%
Sigma-Aldrich
Butyraldehyde, natural, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Butyraldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Propionaldehyde, ≥97%, FG
Supelco
Propionaldehyde, analytical standard
Sigma-Aldrich
Propionaldehyde, reagent grade, 97%
Sigma-Aldrich
Butyraldehyde, ≥99.0%, dry
Supelco
Butyraldehyde, analytical standard, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabilizer
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG
Sigma-Aldrich
Acetaldehyde, natural, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Supelco
Hexanal, analytical standard
Sigma-Aldrich
Acetaldehyde, ≥99%, meets FCC analytical specification
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%