Merck
  • STE20/SPS1-related proline/alanine-rich kinase (SPAK) is critical for sodium reabsorption in isolated, perfused thick ascending limb.
STE20/SPS1-related proline/alanine-rich kinase (SPAK) is critical for sodium reabsorption in isolated, perfused thick ascending limb.
American journal of physiology. Renal physiology (2014-12-06)
Chih-Jen Cheng, Joonho Yoon, Michel Baum, Chou-Long Huang
ABSTRACT
SPAK [STE20 (sterile 20)/SPS1-related proline/alanine-rich kinase] kinase consists of a full-length (FL-) and an alternatively spliced kidney-specific (KS-) isoform. SPAK regulates the NaCl cotransporter (NCC) in the distal convoluted tubule (DCT). The relative abundance and role of FL- vs. KS-SPAK in regulating Na(+)-K(+)-2Cl(-) cotransporter (NKCC2) in thick ascending limb (TAL) are not completely understood. Here, we report that FL-SPAK mRNA was the most abundant in medullary TAL (mTAL), followed by cortical TAL (cTAL) and DCT. KS-SPAK mRNA abundance was relatively lower than FL-SPAK. The ratios of FL-SPAK to KS-SPAK in mTAL, cTAL, and DCT were 12.3, 12.5, and 10.2, respectively. To examine the role of SPAK in the regulation of sodium transport in TAL, we used in vitro microperfusion of mTAL and cTAL isolated from wild-type (WT) and SPAK knockout mice (SPAK-KO) that lack both FL- and KS-SPAK. The rates of sodium absorption in cTAL and mTAL of SPAK-KO mice were 34.5 and 12.5% of WT tubules, respectively. The mRNA levels of related OSR1 kinase and SPAK protease Dnpep in SPAK-KO tubules were not significantly different from WT tubules. We next examined the role of SPAK in the regulation of sodium reabsorption by vasopressin in TAL. Vasopressin increased sodium reabsorption by ∼80% in both mTAL and cTAL from WT mice. While baseline sodium reabsorption was lower in SPAK-KO tubules, vasopressin increased sodium reabsorption over twofold. In conclusion, the combined net effect of SPAK isoforms on sodium reabsorption in TAL is stimulatory. SPAK is not essential for vasopressin stimulation of sodium reabsorption in TAL.
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%