Merck
  • Subconjunctivally applied naïve Tregs support corneal graft survival in baby rats.
Subconjunctivally applied naïve Tregs support corneal graft survival in baby rats.
Molecular vision (2015-01-06)
Antonia Hildebrand, Christian Jarsch, Yvonne Kern, Daniel Böhringer, Thomas Reinhard, Johannes Schwartzkopff
ABSTRACT
Corneal transplantation is the most frequent and successful form of tissue transplantation in adults (<10% rejection). In young children, any corneal opacity should be corrected as early as possible to prevent lifelong visual impairment. However, the corneal graft rejection rate is dramatically increased in infants younger than 12 months of age (up to 85% rejection), and immunosuppressive therapy is particularly challenging in this age group. Regulatory T cells (Tregs) are a well-characterized T cell subpopulation with the potential to prevent autoimmune disorders or transplant rejection. Antigen-specific Tregs were shown to inhibit graft rejection in adult stem cell transplantation. Less is known about the role of naïve Tregs.The purpose of the present study was to elucidate the relevance of naïve Tregs in juvenile corneal transplantation in a baby rat keratoplasty model that reproduces the accelerated rejection in young patients. Counts and inhibitory potential of Tregs were studied in spleens of 3- and 10-week-old rats. Unprimed Tregs (CD4+CD25+) were isolated from the spleens of 10-week-old Lewis rats and systemically or subconjunctivally administered in vivo in allogenic keratoplasty in 3- and 10-week-old Lewis recipient rats. In subconjunctival tissue, transcription was analyzed for induction of transforming-growth-factor beta (TGF-β). In 3-week-old rats, CD4 T cell counts, but not FoxP3 T cell counts were lower than in 10-week-old rats. The Tregs of both age groups had the potential to inhibit T cell activation in vitro. No significant delay in rejection was observed when Tregs were applied systemically before keratoplasty. However, subconjunctival application of Tregs abrogated rejection in 66.7% and 33.3% of the 3- and 10-week-old recipients, respectively. Analysis of the conjunctival tissue revealed a transplantation-induced increase in TGF-β transcription in the 3-week-old rats. Our data suggest that local application of unprimed regulatory T cells may be a therapeutic strategy for preventing corneal graft rejection in young recipients.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%