Merck
Detection of human polyomavirus 7 in human thymic epithelial tumors.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer (2014-12-20)
Dorit Rennspiess, Sreedhar Pujari, Marlies Keijzers, Myrurgia A Abdul-Hamid, Monique Hochstenbag, Anne-Marie Dingemans, Anna Kordelia Kurz, Ernst-Jan Speel, Anke Haugg, Diana V Pastrana, Christopher B Buck, Marc H De Baets, Axel Zur Hausen
ABSTRACT
Although the molecular genetics possibly underlying the pathogenesis of human thymoma have been extensively studied, its etiology remains poorly understood. Because murine polyomavirus consistently induces thymomas in mice, we assessed the presence of the novel human polyomavirus 7 (HPyV7) in human thymic epithelial tumors. HPyV7-DNA Fluorescence in situ hybridization (FISH), DNA-polymerase chain reaction (PCR), and immunohistochemistry (IHC) were performed in 37 thymomas. Of these, 26 were previously diagnosed with myasthenia gravis (MG). In addition, 20 thymic hyperplasias and 20 fetal thymic tissues were tested. HPyV7-FISH revealed specific nuclear hybridization signals within the neoplastic epithelial cells of 23 thymomas (62.2%). With some exceptions, the HPyV7-FISH data correlated with the HPyV7-DNA PCR. By IHC, large T antigen expression of HPyV7 was detected, and double staining confirmed its expression in the neoplastic epithelial cells. Eighteen of the 26 MG-positive and 7 of the 11 MG-negative thymomas were HPyV7-positive. Of the 20 hyperplastic thymi, 40% were HPyV7-positive by PCR as confirmed by FISH and IHC in the follicular lymphocytes. All 20 fetal thymi tested HPyV7-negative. The presence of HPyV7-DNA and large T antigen expression in the majority of thymomas possibly link HPyV7 to human thymomagenesis. Further investigations are needed to elucidate the possible associations of HPyV7 and MG.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade