Merck
  • Gallic acid functions as a TRPA1 antagonist with relevant antinociceptive and antiedematogenic effects in mice.
Gallic acid functions as a TRPA1 antagonist with relevant antinociceptive and antiedematogenic effects in mice.
Naunyn-Schmiedeberg's archives of pharmacology (2014-04-12)
Gabriela Trevisan, Mateus F Rossato, Raquel Tonello, Carin Hoffmeister, Jonatas Z Klafke, Fernanda Rosa, Kelly V Pinheiro, Francielle V Pinheiro, Aline A Boligon, Margareth L Athayde, Juliano Ferreira
ABSTRACT
The transient receptor potential ankyrin 1 (TRPA1) has been identified as a relevant target for the development of novel analgesics. Gallic acid (GA) is a polyphenolic compound commonly found in green tea and various berries and possesses a wide range of biological activities. The goal of this study was to identify GA as a TRPA1 antagonist and observe its antinociceptive effects in different pain models. First, we evaluated the ability of GA to affect cinnamaldehyde-induced calcium influx. Then, we observed the antinociceptive and antiedematogenic effects of GA (3-100 mg/kg) oral administration after the intraplantar (i.pl.) injection of TRPA1 agonists (allyl isothiocyanate, cinnamaldehyde, or hydrogen peroxide-H2O2) in either an inflammatory pain model (carrageenan i.pl. injection) or a neuropathic pain model (chronic constriction injury) in male Swiss mice (25-35 g). GA reduced the calcium influx mediated by TRPA1 activation. Moreover, the oral administration of GA decreased the spontaneous nociception triggered by allyl isothiocyanate, cinnamaldehyde, and H2O2. Carrageenan-induced allodynia and edema were largely reduced by the pretreatment with GA. Moreover, the administration of GA was also capable of decreasing cold and mechanical allodynia in a neuropathic pain model. Finally, GA was absorbed after oral administration and did not produce any detectable side effects. In conclusion, we found that GA is a TRPA1 antagonist with antinociceptive properties in relevant models of clinical pain without detectable side effects, which makes it a good candidate for the treatment of painful conditions.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
SAFC
Sodium chloride solution, 5 M
USP
Capsaicin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Capsaicin, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Allyl isothiocyanate, 95%
Sigma-Aldrich
Cinnamaldehyde, natural, ≥95%, FG
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Allyl isothiocyanate, PESTANAL®, analytical standard
Sigma-Aldrich
Fura 2-AM, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material