Merck
  • Inhibition of stress induced premature senescence in presenilin-1 mutated cells with water soluble Coenzyme Q10.
Inhibition of stress induced premature senescence in presenilin-1 mutated cells with water soluble Coenzyme Q10.
Mitochondrion (2014-07-19)
Dennis Ma, Kyle Stokes, Kevinjeet Mahngar, Danijela Domazet-Damjanov, Marianna Sikorska, Siyaram Pandey
ABSTRACT
A water-soluble formulation of CoQ10 (WS-CoQ10) was shown to stabilize mitochondria and prevent oxidative stress-induced neuronal death. Presenilin-1 (PS-1)-mutated Alzheimer's Disease (AD) fibroblasts (PSAF) were used for studying the effects of PS-1 mutation. PS-1 mutation correlated to increased reactive oxygen species (ROS) production and stress induced premature senescence (SIPS) in PSAF; WS-CoQ10 treatment decreased ROS generation, increased population doublings, and postponed SIPS. Treated PSAF had higher PCNA expression, and lower levels of MnSOD, p21, p16Ink4A, and Rb. WS-CoQ10 caused the resumption of autophagy in PSAF. Thus, WS-CoQ10 as inhibitor of SIPS and ameliorator of autophagy could be an effective prophylactic/therapeutic agent for AD.
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sebacic acid, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Sebacic acid, ≥95.0% (GC)
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%