Merck
  • The aberrant gene-end transcription signal of the matrix M gene of human parainfluenza virus type 3 downregulates fusion F protein expression and the F-specific antibody response in vivo.
The aberrant gene-end transcription signal of the matrix M gene of human parainfluenza virus type 3 downregulates fusion F protein expression and the F-specific antibody response in vivo.
Journal of virology (2015-01-16)
Matthias Lingemann, Sonja Surman, Emérito Amaro-Carambot, Anne Schaap-Nutt, Peter L Collins, Shirin Munir
ABSTRACT
Human parainfluenza virus type 3 (HPIV3), a paramyxovirus, is a major viral cause of severe lower respiratory tract disease in infants and children. The gene-end (GE) transcription signal of the HPIV3 matrix (M) protein gene is identical to those of the nucleoprotein and phosphoprotein genes except that it contains an apparent 8-nucleotide insert. This was associated with an increased synthesis of a readthrough transcript of the M gene and the downstream fusion (F) protein gene. We hypothesized that this insert may function to downregulate the expression of F protein by interfering with termination/reinitiation at the M-F gene junction, thus promoting the production of M-F readthrough mRNA at the expense of monocistronic F mRNA. To test this hypothesis, two similar recombinant HPIV3 viruses from which this insert in the M-GE signal was removed were generated. The M-GE mutants exhibited a reduction in M-F readthrough mRNA and an increase in monocistronic F mRNA. This resulted in a substantial increase in F protein synthesis in infected cells as well as enhanced incorporation of F protein into virions. The efficiency of mutant virus replication was similar to that of wild-type (wt) HPIV3 both in vitro and in vivo. However, the F-protein-specific serum antibody response in hamsters was increased for the mutants compared to wt HPIV3. This study identifies a previously undescribed viral mechanism for attenuating the host adaptive immune response. Repairing the M-GE signal should provide a means to increase the antibody response to a live attenuated HPIV3 vaccine without affecting viral replication and attenuation. The HPIV3 M-GE signal was previously shown to contain an apparent 8-nucleotide insert that was associated with increased synthesis of a readthrough mRNA of the M gene and the downstream F gene. However, whether this had any significant effect on the synthesis of monocistronic F mRNA or F protein, virus replication, virion morphogenesis, and immunogenicity was unknown. Here, we show that the removal of this insert shifts F gene transcription from readthrough M-F mRNA to monocistronic F mRNA. This resulted in a substantial increase in the amount of F protein expressed in the cell and packaged in the virus particle. This did not affect virus replication but increased the F-specific antibody response in hamsters. Thus, in wild-type HPIV3, the aberrant M-GE signal operates a previously undescribed mechanism that reduces the expression of a major neutralization and protective antigen, resulting in reduced immunogenicity. This has implications for the design of live attenuated HPIV3 vaccines; specifically, the antibody response against F can be elevated by "repairing" the M-GE signal to achieve higher-level F antigen expression, with no effect on attenuation.
Product Description
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
MOPS
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free