Merck
Clathrin-mediated endocytosis of gold nanoparticles in vitro.
Anatomical record (Hoboken, N.J. : 2007) (2014-09-23)
Cheng Teng Ng, Florence Mei Ai Tang, Jasmine Jia'en Li, Cynthia Ong, Lanry Lin Yue Yung, Boon Huat Bay
ABSTRACT
Gold nanoparticles (AuNPs) have potential biomedical and scientific applications. In this study, we evaluated the uptake and internalization of FBS-coated 20 nm AuNPs into lung fibroblasts and liver cells by different microscopy techniques. AuNP aggregates were observed inside MRC5 lung fibroblasts and Chang liver cells under light microscopy, especially after enhancement with automegallography. Clusters of AuNPs were observed to be adsorbed on the cell surface by scanning electron microscopy. Ultrathin sections showed that AuNPs were mainly enclosed within cytoplasmic vesicles when viewed under transmission electron microscopy. We also investigated the mechanism of uptake for AuNPs, using endocytosis inhibitors and quantification of Au with inductively coupled plasma mass spectrometry. Cells treated with concanavalin A and chlorpromazine showed significant decrease of Au uptake in MRC5 lung fibroblasts and Chang liver cells, respectively, implying that the uptake of AuNPs was facilitated by clathrin-mediated endocytosis. It would therefore appear that uptake of 20 nm AuNPs in both cell types with different tissues of origin, was dependent upon clathrin-mediated endocytosis.
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O