Merck
A20-deficient mast cells exacerbate inflammatory responses in vivo.
PLoS biology (2014-01-24)
Klaus Heger, Kaat Fierens, J Christoph Vahl, Attila Aszodi, Katrin Peschke, Dominik Schenten, Hamida Hammad, Rudi Beyaert, Dieter Saur, Geert van Loo, Axel Roers, Bart N Lambrecht, Mirjam Kool, Marc Schmidt-Supprian
ABSTRACT
Mast cells are implicated in the pathogenesis of inflammatory and autoimmune diseases. However, this notion based on studies in mast cell-deficient mice is controversial. We therefore established an in vivo model for hyperactive mast cells by specifically ablating the NF-κB negative feedback regulator A20. While A20 deficiency did not affect mast cell degranulation, it resulted in amplified pro-inflammatory responses downstream of IgE/FcεRI, TLRs, IL-1R, and IL-33R. As a consequence house dust mite- and IL-33-driven lung inflammation, late phase cutaneous anaphylaxis, and collagen-induced arthritis were aggravated, in contrast to experimental autoimmune encephalomyelitis and immediate anaphylaxis. Our results provide in vivo evidence that hyperactive mast cells can exacerbate inflammatory disorders and define diseases that might benefit from therapeutic intervention with mast cell function.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Aluminum hydroxide, reagent grade
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, ≥99%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Anti -Dinitrophenyl antibody, Mouse monoclonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Calcium chloride
Supelco
1-Fluoro-2,4-dinitrobenzene, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Supelco
4-Hydroxytamoxifen, (E) and (Z) isomers (50:50), analytical standard
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
1-Fluoro-2,4-dinitrobenzene, purum p.a., ≥98.0% (GC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Calcium chloride, Vetec, reagent grade, 96%