Merck
  • Sex differences in the effects of adolescent social deprivation on alcohol consumption in μ-opioid receptor knockout mice.
Sex differences in the effects of adolescent social deprivation on alcohol consumption in μ-opioid receptor knockout mice.
Psychopharmacology (2014-11-05)
Yuki Moriya, Yoshiyuki Kasahara, F Scott Hall, Yasufumi Sakakibara, George R Uhl, Hiroaki Tomita, Ichiro Sora
ABSTRACT
Evidence based on clinical and experimental animal studies indicates that adolescent social deprivation influences alcohol consumption in a sex-dependent manner, perhaps by influencing stress responses. However, the mechanisms underlying the interaction between these phenomena remain to be elucidated. Since the μ-opioid receptor (MOP) has been reported to have key roles in social stress responses as well as the reinforcing/addictive effects of ethanol, MOP is a candidate molecule that may link adolescent social deprivation and subsequent alterations in alcohol consumption. To evaluate the involvement of MOP and social isolation-induced changes in alcohol consumption, as well as the effect of sex differences on responses to social isolation, alcohol consumption was assessed using a two-bottle home-cage consumption procedure (8 % ethanol vs. water) in MOP knockout (MOP-KO) and wild type (WT) mice of both sexes exposed to adolescent social deprivation or reared socially. Isolation rearing had no effects upon alcohol consumption of WT mice, whereas it significantly altered alcohol consumption in both male and female MOP-KO mice. Interestingly, social isolation affected ethanol consumption differently in male and female mice. Ethanol consumption was increased in male MOP-KO mice, but decreased in female MOP-KO mice, by isolation rearing. These results indicate that disturbances of MOP function influence the effects of isolation rearing on ethanol consumption in a sex-dependent manner. Consequently, this suggests the possibility that genetic variation that influences MOP function may have differential roles in alcoholism in men and women, and alcoholism treatments that target MOP function may be differentially effective in males and females.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications