Merck
  • Serotonin and sudden death: differential effects of serotonergic drugs on seizure-induced respiratory arrest in DBA/1 mice.
Serotonin and sudden death: differential effects of serotonergic drugs on seizure-induced respiratory arrest in DBA/1 mice.
Epilepsy & behavior : E&B (2014-07-30)
Carl L Faingold, Srinivasa P Kommajosyula, X Long, Kristin Plath, Marcus Randall
ABSTRACT
In the DBA/1 mouse model of sudden unexpected death in epilepsy (SUDEP), administration of a selective serotonin (5-HT) reuptake inhibitor (SSRI), fluvoxamine, completely suppressed seizure-induced respiratory arrest (S-IRA) at 30 min after administration (i.p.) in a dose-related manner without blocking audiogenic seizures (AGSz), but another SSRI, paroxetine, reduced S-IRA but with a delayed (24 h) onset and significant toxicity. A serotonin-norepinephrine reuptake inhibitor, venlafaxine, reduced S-IRA incidence, but higher doses were ineffective. A selective 5-HT7 agonist, AS-19, was totally ineffective in reducing S-IRA. In developing DBA/1 mice that had not previously experienced AGSz, administration of a nonselective 5-HT antagonist, cyproheptadine, induced a significantly greater incidence of S-IRA than that of saline. This study confirms that certain drugs that enhance the activation of 5-HT receptors are able to prevent S-IRA, but not all serotonergic drugs are equally effective, which may be relevant to the potential use of these drugs for SUDEP prevention. Serotonergic antagonists may be problematic in patients with epilepsy.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)