Merck
  • Preparation and evaluation of benzyl methacrylate monoliths for capillary chromatography.
Preparation and evaluation of benzyl methacrylate monoliths for capillary chromatography.
Journal of chromatographic science (2013-02-21)
Ahmad Aqel, Zeid A ALOthman, Kareem Yusuf, A Yacine Badjah-Hadj-Ahmed, Abdulrahman A Alwarthan
ABSTRACT
This paper describes the comprehensive fabrication of monolithic materials for use as stationary phases in capillary liquid chromatography. Several columns were synthesized in the confines of 320 µm i.d. fused-silica capillaries by single-step in situ copolymerization of benzyl methacrylate and ethylene dimethacrylate (EDMA). The polymerization procedure was optimized by varying the reaction time within the range of 0.5-20 h, and by changing the composition contents of the polymeric mixture. The EDMA content showed a predominant influence on the characteristics of the columns and hence, on their chromatographic properties. The optimum value of the thermal initiator corresponded to 5 mg/mL. Changes of the porous, hydrodynamic properties and morphology of the prepared columns were thoroughly investigated and characterized. Different solvents were used as the mobile phase to demonstrate that the resulting monoliths exhibited good permeability and mechanical stability, whereas swelling and shrinking behaviors were observed and discussed. The efficiency and performance toward different sets of analytes were obtained; mixtures of aromatic hydrocarbons and phenolic compounds were successfully separated and evaluated, and adding tetrahydrofuran to the mobile phase showed improvement in both resolution and peak shapes. The characteristics of the columns were also checked in terms of repeatability and reproducibility.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
4-Aminophenol (Acetaminophen Related Compound K) (Paracetamol Impurity K), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 79-81°C, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
Sigma-Aldrich
3-(Trimethoxysilyl)propyl methacrylate, 98%
Supelco
4-Aminophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, ≥99.0% (GC)
Supelco
Benzene, analytical standard
Supelco
Toluene, analytical standard
Sigma-Aldrich
2-Naphthol, BioXtra, ≥99.0% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
4-Aminophenol, ≥98%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
2-Naphthol, 98%
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Supelco
Hexane, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis