Merck
  • Limoniastrum guyonianum methanol extract induces immunomodulatory and anti-inflammatory effects by activating cellular anti-oxidant activity.
Limoniastrum guyonianum methanol extract induces immunomodulatory and anti-inflammatory effects by activating cellular anti-oxidant activity.
Drug and chemical toxicology (2014-04-15)
Mounira Krifa, Nadia Mustapha, Zied Ghedira, Kamel Ghedira, Antonio Pizzi, Leila Chekir-Ghedira
ABSTRACT
Evaluation of the immunomodulatory activity of plant extracts is an interesting and growing area of research. In this study, effects of a methanolic extract of Limoniastrum guyonianum stems (M extract) on mice immune cell function in vitro were investigated. These studies showed that mitogen-induced lymphocyte proliferation was dose-dependently inhibited by the extract. Further, the lectin-induced response appeared to be more sensitive to the suppressive effects of the extract than were LPS-stimulated responses. In studies to assess any potential effects of extract on innate immunity, the results showed that the extract significantly enhanced the killing activity of isolated NK cells. In addition, studies here demonstrated that the extract could enhance lysosomal enzyme activity and inhibit nitrite oxide (NO) production by murine peritoneal macrophages ex vivo, suggesting a potential anti-inflammatory effect in situ. The anti-inflammatory activity was concomitant with the cellular anti-oxidant effect in macrophages and splenocytes.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Tannic acid, Source: Chinese natural gall nuts
Sigma-Aldrich
Tannic acid, ACS reagent
Sigma-Aldrich
Tannic acid
SAFC
Sodium chloride solution, 5 M
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%