Merck
  • (6aR)-11-amino-N-propyl-noraporphine, a new dopamine D2 and serotonin 5-HT1A dual agonist, elicits potent antiparkinsonian action and attenuates levodopa-induced dyskinesia in a 6-OHDA-lesioned rat model of Parkinson's disease.
(6aR)-11-amino-N-propyl-noraporphine, a new dopamine D2 and serotonin 5-HT1A dual agonist, elicits potent antiparkinsonian action and attenuates levodopa-induced dyskinesia in a 6-OHDA-lesioned rat model of Parkinson's disease.
Pharmacology, biochemistry, and behavior (2014-06-24)
Rui Zhao, Weijian Lu, Xing Fang, Lin Guo, Zhi Yang, Na Ye, Jiahao Zhao, Zhili Liu, Jia Jia, Longtai Zheng, Bin Zhao, Ao Zhang, Xuechu Zhen
ABSTRACT
Parkinson's disease (PD) drug therapy remains a challenge. Dual modulation of dopamine and 5-HT receptors has emerged as a promising approach in anti-PD drug development. Taking advantage of the newly discovered aporphine analogue(s), (6aR)-11-amino-N-propyl-noraporphine (SOMCL-171), which exhibited dual D2/5-HT1A receptor agonistic activity, we studied the effects of the compound on levodopa-induced dyskinesia (LID) in a PD animal model. The results demonstrated that SOMCL-171 elicited a potent anti-PD effect in a 6-OHDA-lesioned rat model. Chronic use of SOMCL-171 reduced LID without compromising the antiparkinsonian efficacy. Furthermore, we found that the antidyskinesia effect of SOMCL-171 is associated with its 5-HT1A agonistic activity and the up-regulation of the striatal 5-HT1A receptor. The present data indicated that chronic SOMCL-171 alone produced potent antiparkinsonian effects with weak dyskinesia, compared with that of levodopa. In addition, chronic SOMCL-171 application attenuated the development of levodopa-induced LID at no expense to the antiparkinsonian efficacy. Taken together, our data suggested that dual modulation of D2/5-HT1A receptors may provide a novel approach for drug development in PD and LID.
Sigma-Aldrich
R-(−)-Apomorphine hydrochloride hemihydrate, calcined, ≥98% (with NaOH, titration)
Apomorphine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
USP
Desipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard