Merck
  • Hepatic oxidative status and metal homeostasis disturbance of 2-hydroxylated dioxin in ICR mice.
Hepatic oxidative status and metal homeostasis disturbance of 2-hydroxylated dioxin in ICR mice.
Environmental toxicology and pharmacology (2014-12-03)
Xuesheng Zhang, Li Qin, Dinglong Li, Zhongbo Wei, Zunyao Wang
ABSTRACT
In the present study, the toxic effects of the oral exposure of 2-hydroxylated dioxin (2-HODD) in ICR male mice were examined. The mice were administered different doses (0.2, 2.0 and 20.0mg/kg) of 2-HODD. After 14 days of exposure, the oxidative stress (OS) indicator levels and the essential metal concentrations in the mouse livers were determined. The results showed that the superoxide dismutase (SOD) and the glutathione peroxidase (GPx) activities were increased in the 0.2mg/kg group, whereas they were significantly decreased in the 2.0 and 20.0mg/kg groups. Decreases in the catalase (CAT) activity and the glutathione (GSH) levels, accompanied by increases in the malondialdehyde (MDA) contents, were recorded in all of the 2-HODD-treated groups. The hepatic iron, copper and zinc levels increased in all of the 2-HODD-treated groups. The histological examination of the livers demonstrated swollen cells and inflammation. Dose-dependent changes in both the OS indicators and the hepatic metal levels were observed. In conclusion, a single low dose of 2-HODD significantly perturbed the hepatic OS status and metals homeostasis in the mice.
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Cesium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Copper(I) iodide, purum, ≥99.5%
Sigma-Aldrich
Cesium carbonate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
1,4-Diaminobutane, 99%
Sigma-Aldrich
Cesium carbonate, 99.9% trace metals basis
Sigma-Aldrich
Cesium carbonate, puriss. p.a., ≥99.0%
Sigma-Aldrich
Copper(I) iodide, 99.999% trace metals basis
Sigma-Aldrich
Cesium carbonate, 99.95% trace metals basis
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Supelco
4,4′-Diaminodiphenylmethane, analytical standard
Supelco
Putrescine, analytical standard
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Copper(I) iodide, anhydrous, 99.995% trace metals basis
Sigma-Aldrich
Copper(I) iodide, 98%
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%