Merck
  • A multiresidue method for the determination of selected endocrine disrupting chemicals in human breast milk based on a simple extraction procedure.
A multiresidue method for the determination of selected endocrine disrupting chemicals in human breast milk based on a simple extraction procedure.
Talanta (2014-08-28)
R Rodríguez-Gómez, I Jiménez-Díaz, A Zafra-Gómez, O Ballesteros, A Navalón
ABSTRACT
In recent decades, in parallel to industrial development, a large amount of new chemicals have emerged that are able to produce disorders in human endocrine system. These groups of substances, so-called endocrine disrupting chemicals (EDCs), include many families of compounds, such as parabens, benzophenone-UV filters and bisphenols. Given the demonstrated biological activity of those compounds, it is necessary to develop new analytical procedures to evaluate the exposure with the final objective of establishing, in an accurate way, relationships between EDCs concentrations and the harmful health effects observed in population. In the present work, a method based on a simplified sample treatment involving steps of precipitation, evaporation and clean-up of the extracts with C18 followed by ultrahigh performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) analysis for the determination of bisphenol A and its chlorinated derivatives (monochloro-, dichloro-, trichloro- and tetrachlorobisphenol A), parabens (methyl-, ethyl-, propyl- and butylparaben) and benzophenone-UV filters (benzophenone -1,-2, -3, -6, -8 and 4-hydroxybenzophenone) in human breast milk samples is proposed and validated. The limits of detections found ranged from 0.02 to 0.05 ng mL(-1). The method was validated using matrix-matched standard calibration followed by a recovery assay with spiked samples. Recovery rates ranged from 91% to 110% and the precision (evaluated as relative standard deviation) was lower than 15% for all compounds, being within the acceptable limits for the selected bioanalytical method validation guide. The method was satisfactorily applied for the determination of these compounds in human breast milk samples collected from 10 randomly selected women.
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Deuterium, 99.9 atom % D
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Deuterium, 99.8 atom % D
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Phosphotungstic acid hydrate, 99.995% trace metals basis (Purity excludes up to 300 ppm Si)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Deuterium, 99.96 atom % D
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Phosphotungstic acid hydrate, reagent grade