Merck
A profluorescent ratiometric probe for intracellular pH imaging.
Talanta (2014-10-05)
Qingqing Zhang, Ming Zhou
ABSTRACT
A ratiometric pH probe composed of a fluorescein moiety and an ionic near-infrared-emitting phosphorescent cyclometalated iridium(III) complex bis(6-(benzo[b]thien-2-yl)phenanthridinato)(4-(3-carboxypropyl)-4'-methyl-2,2'-bipyridine)iridium(III) was synthesized. With good cell permeability, the probe demonstrated a linear ratiometric response to the pH variation in the physiological range in HeLa cell assay.
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, ≥99%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ReagentPlus®, 99%
Sigma-Aldrich
Di-tert-butyl dicarbonate, ≥98.0% (GC)
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in methylene chloride
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 2 M in THF
Sigma-Aldrich
Di-tert-butyl dicarbonate solution, 1.0 M in THF
Sigma-Aldrich
Hexamethylenediamine, 98%
Sigma-Aldrich
Hexamethylenediamine, technical grade, 70%