Merck
  • Effects of the addition of dimer acid alkyl esters on the properties of ethyl cellulose.
Effects of the addition of dimer acid alkyl esters on the properties of ethyl cellulose.
Carbohydrate polymers (2015-02-11)
Sangjun Lee, Kwang-Hwan Ko, Jihoon Shin, Nam-Kyun Kim, Young-Wun Kim, Joon-Seop Kim
ABSTRACT
In this study, we synthesized dimer acid (DA) esters, having short to long alkyl chains, (DA-Cn) by the Diels-Alder reaction and subsequent esterification reaction of fatty acids that were prepared by the hydrolysis of waste vegetable oil. It was found that the DA-Cn were thermally more stable than common petroleum-based plasticizer DOP. When the DOP, DA, or DA-Cn with short alkyl chains were added to ethyl cellulose (EC), the optical clarity and SEM images of the samples showed their good miscibility with those additives in a micro-scale. It was also found that the rubbery modulus of the EC decreased with increasing amount of additives; the type of the additives did not affect the rates of the decrease in the rubbery modulus. The main transition temperatures of the EC containing either DA or DA-C1 or DA-C4 decreased with increasing amounts of those additives and were comparable to that of the DOP-containing EC. The above findings suggested that the DA and its esters with short alkyl chains could act as effective plasticizer and, thus, could be used instead of the DOP. In addition, the results obtained from tensile testing and leaching experiments implied that the DA might be better plasticizer than the DA-C1 and DA-C4, at least in some cases, because of hydrogen-bonding with the EC.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Supelco
Methanol, analytical standard
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99%, FG
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Supelco
2-Ethyl-1-hexanol, analytical standard
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
1-Butanol, analytical standard
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Ethyl-1-hexanol, ≥99.6%
Sigma-Aldrich
2-Ethyl-1-hexanol, puriss., ≥99.0% (GC)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%