Merck
  • EGb-761 prevents ultraviolet B-induced photoaging via inactivation of mitogen-activated protein kinases and proinflammatory cytokine expression.
EGb-761 prevents ultraviolet B-induced photoaging via inactivation of mitogen-activated protein kinases and proinflammatory cytokine expression.
Journal of dermatological science (2014-05-08)
Chih-Chiang Chen, An-Na Chiang, Han-Nan Liu, Yun-Ting Chang
ABSTRACT
EGb-761 is an antioxidant and anticarcinogen; however, its role as a photoprotector remains unknown. To determine whether EGb-761 photoprotects human dermal fibroblasts and BALB/c mice skin against ultraviolet B (UVB) light irradiation. To simulate chronic photodamage, shaved BALB/c mice were exposed to UVB irradiation (90mJ/cm(2)) thrice weekly for 3 months. EGb-761 (2mg/cm(2)) was topically applied 1h before irradiation to evaluate its effect. The mechanisms by which EGb-761 protects the skin from photodamage were evaluated by immunohistochemical analysis, enzyme-linked immunosorbent assay (ELISA), and Western blotting. In BALB/c mice, the signs of photoaging or photodamage, such as coarse wrinkle formation, epidermal hyperplasia, and elastic fiber degeneration, markedly reduced with the topical application of EGb-761. Western blot and ELISA revealed that the activation of MMP-1 in cultured fibroblasts markedly diminished after pretreatment with EGb-761. In addition, EGb-761 inhibited UVB-induced overexpression by the fibroblasts of the proinflammatory cytokines, such as interleukin (IL)-1α, IL-1β, IL-6, and tumor necrosis factor-α. The phosphorylation of the mitogen-activated protein kinase (MAPK) signal transduction pathway components, including extracellular signal-regulated kinase, C-Jun N-terminal kinase, and p38, which are induced by UV irradiation, was significantly inhibited in vivo and in vitro. EGb-761 also diminished the generation of UVB-induced reactive oxygen species (ROS). EGb-761 photoprotects mice and cultured fibroblasts, inhibits the UVB-induced phosphorylation of MAPK pathway components, and reduces the expression of the proinflammatory cytokines by suppressing ROS generation. Thus, topically applied EGb-761 may be a promising photoprotective agent.
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
SP600125, ≥98% (HPLC)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard