Merck
  • Electrochemical tuning of MoS2 nanoparticles on three-dimensional substrate for efficient hydrogen evolution.
Electrochemical tuning of MoS2 nanoparticles on three-dimensional substrate for efficient hydrogen evolution.
ACS nano (2014-04-11)
Haotian Wang, Zhiyi Lu, Desheng Kong, Jie Sun, Thomas M Hymel, Yi Cui
ABSTRACT
Molybdenum disulfide (MoS2) with the two-dimensional layered structure has been widely studied as an advanced catalyst for hydrogen evolution reaction (HER). Intercalating guest species into the van der Waals gaps of MoS2 has been demonstrated as an effective approach to tune the electronic structure and consequently improve the HER catalytic activity. In this work, by constructing nanostructured MoS2 particles with largely exposed edge sites on the three-dimensional substrate and subsequently conducting Li electrochemical intercalation and exfoliation processes, an ultrahigh HER performance with 200 mA/cm(2) cathodic current density at only 200 mV overpotential is achieved. We propose that both the high surface area nanostructure and the 2H semiconducting to 1T metallic phase transition of MoS2 are responsible for the outstanding catalytic activity. Electrochemical stability test further confirms the long-term operation of the catalyst.
Product Description
Supelco
Sulfur, PESTANAL®, analytical standard
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Diethyl carbonate, 99%
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis