USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
1-Hydroxy-7-azabenzotriazole solution, ~0.6 M in DMF
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Pricing and availability is not currently available.